Assay Details
Gene Symbol: Ttc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503291.2 | XP_003503339.1 | 4 - 5 | 627 | 64 | |
XM_007643894.1 | XP_007642084.1 | 4 - 5 | 649 | 64 | ||
XM_007643895.1 | XP_007642085.1 | 4 - 5 | 646 | 64 | ||
GenBank mRNA | JI862433.1 | - | 3 - 4 | 441 | 64 | |
JP050950.1 | - | 4 - 5 | 486 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
