Assay Details
Gene Symbol: Ccnjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642831.1 | XP_007641021.1 | 6 - 7 | 919 | 66 | |
GenBank mRNA | JI889684.1 | - | 2 - 3 | 390 | 66 | |
JP044229.1 | - | 2 - 3 | 390 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
