Assay Details
Gene Symbol: Prkrir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647655.1 | XP_007645845.1 | 1 | 98 | 74 | |
GenBank mRNA | JI894525.1 | - | 1 - 2 | 124 | 74 | |
JP051874.1 | - | 1 - 2 | 124 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
