Assay Details
Gene Symbol: Wash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508388.2 | XP_003508436.2 | 6 - 7 | 858 | 60 | |
GenBank mRNA | JI893143.1 | - | 5 - 6 | 680 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
