Assay Details
| Gene Symbol: Mdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501192.2 | XP_003501240.1 | 8 - 9 | 775 | 85 | |
| XM_003501193.2 | XP_003501241.1 | 7 - 8 | 772 | 85 | ||
| XM_007641884.1 | XP_007640074.1 | 8 - 9 | 775 | 85 | ||
| XM_007641885.1 | XP_007640075.1 | 8 - 9 | 714 | 85 | ||
| XM_007641886.1 | XP_007640076.1 | 8 - 9 | 541 | 85 | ||
| XM_007641887.1 | XP_007640077.1 | 7 - 8 | 698 | 85 | ||
| XM_007641888.1 | XP_007640078.1 | 7 - 8 | 637 | 85 | ||
| XM_007641889.1 | XP_007640079.1 | 7 - 8 | 772 | 85 | ||
| XM_007641890.1 | XP_007640080.1 | 7 - 8 | 772 | 85 | ||
| XM_007641891.1 | XP_007640081.1 | 8 - 9 | 775 | 85 | ||
| XM_007641892.1 | XP_007640082.1 | 8 - 9 | 775 | 85 | ||
More Information
Set Membership: |
|

Mdm4
Genomic Map