Assay Details
| Gene Symbol: Atp5s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511591.2 | XP_003511639.1 | 3 - 4 | 390 | 115 | |
| GenBank mRNA | JI863125.1 | - | 3 - 4 | 371 | 115 | |
More Information
Set Membership: |
|

Atp5s
Genomic Map