Assay Details
| Gene Symbol: Sap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651253.1 | XP_007649443.1 | 1 - 2 | 310 | 87 | |
| XM_007651254.1 | XP_007649444.1 | 1 - 2 | 310 | 87 | ||
More Information
Set Membership: |
|

Sap25
Genomic Map