Assay Details
| Gene Symbol: Polr2j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500955.2 | XP_003501003.1 | 1 - 2 | 127 | 69 | |
| GenBank mRNA | JI896822.1 | - | 1 - 2 | 129 | 69 | |
More Information
Set Membership: |
|

Polr2j
Genomic Map