Assay Details
| Gene Symbol: Gpr133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653651.1 | XP_007651841.1 | 11 - 12 | 1736 | 95 | |
| XM_007653652.1 | XP_007651842.1 | 10 - 11 | 1640 | 95 | ||
| GenBank mRNA | JI862646.1 | - | 7 - 8 | 955 | 95 | |
| JP046789.1 | - | 11 - 12 | 1233 | 95 | ||
More Information
Set Membership: |
|

Gpr133
Genomic Map