Assay Details
| Gene Symbol: Gpr133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653651.1 | XP_007651841.1 | 20 - 21 | 2702 | 105 | |
| XM_007653652.1 | XP_007651842.1 | 19 - 20 | 2606 | 105 | ||
More Information
Set Membership: |
|

Gpr133
Genomic Map