Assay Details
| Gene Symbol: Gpr133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653651.1 | XP_007651841.1 | 2 - 3 | 607 | 83 | |
| XM_007653652.1 | XP_007651842.1 | 2 - 3 | 607 | 83 | ||
| GenBank mRNA | JP046789.1 | - | 2 - 3 | 104 | 83 | |
More Information
Set Membership: |
|

Gpr133
Genomic Map