Assay Details
| Gene Symbol: Gpr133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653651.1 | XP_007651841.1 | 8 - 9 | 1403 | 67 | |
| XM_007653652.1 | XP_007651842.1 | 7 - 8 | 1307 | 67 | ||
| GenBank mRNA | JI862646.1 | - | 4 - 5 | 622 | 67 | |
| JP046789.1 | - | 8 - 9 | 900 | 67 | ||
More Information
Set Membership: |
|

Gpr133
Genomic Map