Assay Details
| Gene Symbol: Gpr133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653651.1 | XP_007651841.1 | 9 - 10 | 1558 | 61 | |
| XM_007653652.1 | XP_007651842.1 | 8 - 9 | 1462 | 61 | ||
| GenBank mRNA | JI862646.1 | - | 5 - 6 | 777 | 61 | |
| JP046789.1 | - | 9 - 10 | 1055 | 61 | ||
More Information
Set Membership: |
|

Gpr133
Genomic Map