Assay Details
Gene Symbol: LOC100754898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646200.1 | XP_007644390.1 | 1 - 2 | 164 | 140 | |
XM_007646201.1 | XP_007644391.1 | 1 - 2 | 164 | 140 | ||
GenBank mRNA | JI889978.1 | - | 2 - 3 | 310 | 140 | |
JP042359.1 | - | 2 - 3 | 310 | 140 |

More Information
Set Membership: |
![]() ![]() |
