Assay Details
Gene Symbol: LOC100754898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646200.1 | XP_007644390.1 | 3 - 4 | 432 | 85 | |
GenBank mRNA | JI889978.1 | - | 4 - 5 | 578 | 85 | |
JP042359.1 | - | 4 - 5 | 578 | 85 |
