Assay Details
Gene Symbol: LOC100754898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646200.1 | XP_007644390.1 | 5 - 6 | 641 | 78 | |
GenBank mRNA | JI889978.1 | - | 6 - 7 | 787 | 78 | |
JP042359.1 | - | 6 - 7 | 787 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
