Assay Details
| Gene Symbol: Tfam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007639316.1 | XP_007637506.1 | 2 - 3 | 261 | 66 | |
| GenBank mRNA | JI880363.1 | - | 2 - 3 | 261 | 66 | |
More Information
Set Membership: |
|

Tfam
Genomic Map