Assay Details
| Gene Symbol: Chp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498067.2 | XP_003498115.1 | 2 - 3 | 180 | 61 | |
| XM_007622340.1 | XP_007620530.1 | 2 - 3 | 371 | 61 | ||
| GenBank mRNA | JI881985.1 | - | 1 - 2 | 172 | 61 | |
| JP055697.1 | - | 1 - 2 | 172 | 61 | ||
More Information
Set Membership: |
|

Chp1
Genomic Map