Assay Details
| Gene Symbol: Ckap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503303.2 | XP_003503351.1 | 4 - 5 | 1088 | 62 | |
| XM_007643904.1 | XP_007642094.1 | 4 - 5 | 968 | 62 | ||
| XM_007643905.1 | XP_007642095.1 | 3 - 4 | 981 | 62 | ||
| GenBank mRNA | JI866631.1 | - | 2 - 3 | 847 | 62 | |
| JP039951.1 | - | 2 - 3 | 844 | 62 | ||
More Information
Set Membership: |
|

Ckap2
Genomic Map