Assay Details
| Gene Symbol: Josd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509466.2 | XP_003509514.1 | 1 - 2 | 696 | 67 | |
| XM_007650057.1 | XP_007648247.1 | 1 - 2 | 696 | 67 | ||
| GenBank mRNA | JI863759.1 | - | 1 - 2 | 378 | 67 | |
| JP048335.1 | - | 1 - 2 | 378 | 67 | ||
More Information
Set Membership: |
|

Josd1
Genomic Map