Assay Details
Gene Symbol: Mrps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509506.2 | XP_003509554.1 | 2 - 3 | 425 | 93 | |
GenBank mRNA | JP061858.1 | - | 2 - 3 | 403 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
