Assay Details
| Gene Symbol: Gamt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502613.2 | XP_003502661.2 | 1 - 2 | 181 | 58 | |
| GenBank mRNA | JI885843.1 | - | 1 - 2 | 181 | 58 | |
| JP060665.1 | - | 1 - 2 | 181 | 58 | ||
More Information
Set Membership: |
|

Gamt
Genomic Map