Assay Details
| Gene Symbol: Atp6v1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503447.2 | XP_003503495.1 | 1 - 2 | 205 | 68 | |
| GenBank mRNA | JI885919.1 | - | 1 - 2 | 183 | 68 | |
| JP039966.1 | - | 1 - 2 | 183 | 68 | ||
More Information
Set Membership: |
|

Atp6v1g1
Genomic Map