Assay Details
Gene Symbol: Ppm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512313.2 | XP_003512361.2 | 2 - 3 | 882 | 80 | |
XM_007652828.1 | XP_007651018.1 | 2 - 3 | 918 | 80 | ||
GenBank mRNA | JI895092.1 | - | 1 - 2 | 856 | 80 | |
JP058612.1 | - | 1 - 2 | 856 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
