Assay Details
| Gene Symbol: Det1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514273.2 | XP_003514321.1 | 4 - 5 | 1561 | 68 | |
| XM_007654442.1 | XP_007652632.1 | 5 - 6 | 1696 | 68 | ||
| GenBank mRNA | JI888056.1 | - | 2 - 3 | 381 | 68 | |
| JP053813.1 | - | 2 - 3 | 381 | 68 | ||
More Information
Set Membership: |
|

Det1
Genomic Map