Assay Details
Gene Symbol: Naf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651365.1 | XP_007649555.1 | 7 - 8 | 768 | 91 | |
GenBank mRNA | JI877121.1 | - | 6 - 7 | 570 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
