Assay Details
| Gene Symbol: Pdzk1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497397.2 | XP_003497445.2 | 2 - 3 | 257 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdzk1ip1| Gene Symbol: Pdzk1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497397.2 | XP_003497445.2 | 2 - 3 | 257 | 62 | |
Set Membership: |
|