Assay Details
| Gene Symbol: Kcnk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504569.2 | XP_003504617.1 | 1 - 2 | 336 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk15| Gene Symbol: Kcnk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504569.2 | XP_003504617.1 | 1 - 2 | 336 | 63 | |
Set Membership: |
|