Assay Details
Gene Symbol: Dnttip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513756.2 | XP_003513804.1 | 1 - 2 | 114 | 75 | |
GenBank mRNA | JI893431.1 | - | 1 - 2 | 195 | 75 | |
JP057327.1 | - | 1 - 2 | 195 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
