Assay Details
Gene Symbol: Tmem138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514325.2 | XP_003514373.1 | 3 - 4 | 543 | 65 | |
XM_007654487.1 | XP_007652677.1 | 3 - 4 | 751 | 65 | ||
XM_007654488.1 | XP_007652678.1 | 3 - 4 | 872 | 65 | ||
GenBank mRNA | JI876080.1 | - | 3 - 4 | 487 | 65 | |
JP064257.1 | - | 3 - 4 | 550 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
