Assay Details
Gene Symbol: Urgcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645257.1 | XP_007643447.1 | 4 - 5 | 245 | 69 | |
GenBank mRNA | JI878700.1 | - | 6 - 7 | 335 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
