Assay Details
| Gene Symbol: Rpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503554.2 | XP_003503602.1 | 7 - 8 | 1314 | 64 | |
| GenBank mRNA | JI892471.1 | - | 7 - 8 | 1200 | 64 | |
| JP065831.1 | - | 7 - 8 | 1200 | 64 | ||
More Information
Set Membership: |
|

Rpn1
Genomic Map