Assay Details
| Gene Symbol: Jkamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512798.2 | XP_003512846.1 | 5 - 6 | 791 | 81 | |
| GenBank mRNA | JI871706.1 | - | 5 - 6 | 791 | 81 | |
| JP042145.1 | - | 5 - 6 | 791 | 81 | ||
More Information
Set Membership: |
|

Jkamp
Genomic Map