Assay Details
Gene Symbol: Acbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647794.1 | XP_007645984.1 | 4 - 5 | 841 | 58 | |
GenBank mRNA | JP055649.1 | - | 3 - 4 | 415 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
