Assay Details
Gene Symbol: LOC103158713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007607747.1 | XP_007605937.1 | 2 - 3 | 219 | 121 | |
GenBank mRNA | JI893496.1 | - | 1 - 2 | 80 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
