Assay Details
| Gene Symbol: Fxyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644828.1 | XP_007643018.1 | 4 - 5 | 170 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fxyd4| Gene Symbol: Fxyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644828.1 | XP_007643018.1 | 4 - 5 | 170 | 59 | |
Set Membership: |
|