Assay Details
Gene Symbol: Gch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500048.2 | XP_003500096.1 | 2 - 3 | 614 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
