Assay Details
Gene Symbol: LOC100764246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500358.2 | XP_003500406.1 | 2 - 3 | 1145 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100764246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500358.2 | XP_003500406.1 | 2 - 3 | 1145 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() |