Assay Details
Gene Symbol: Triqk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509325.2 | XP_003509373.1 | 3 - 4 | 312 | 83 | |
XM_007649908.1 | XP_007648098.1 | 4 - 5 | 420 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
