Assay Details
Gene Symbol: Nrsn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511496.2 | XP_003511544.2 | 1 - 2 | 565 | 84 | |
XM_007652038.1 | XP_007650228.1 | 3 - 4 | 486 | 84 | ||
XM_007652039.1 | XP_007650229.1 | 4 - 5 | 609 | 84 | ||
XM_007652041.1 | XP_007650231.1 | 3 - 4 | 523 | 84 | ||
XM_007652042.1 | XP_007650232.1 | 4 - 5 | 632 | 84 | ||
XM_007652043.1 | XP_007650233.1 | 2 - 3 | 446 | 84 | ||
XM_007652044.1 | XP_007650234.1 | 2 - 3 | 659 | 84 | ||
XM_007652045.1 | XP_007650235.1 | 3 - 4 | 381 | 84 | ||
XM_007652047.1 | XP_007650237.1 | 3 - 4 | 482 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
