Assay Details
| Gene Symbol: Rnf114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643095.1 | XP_007641285.1 | 2 - 3 | 411 | 60 | |
| GenBank mRNA | JI863235.1 | - | 2 - 3 | 411 | 60 | |
More Information
Set Membership: |
|

Rnf114
Genomic Map