Assay Details
Gene Symbol: Ypel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496568.2 | XP_003496616.1 | 2 - 3 | 188 | 70 | |
GenBank mRNA | JI880511.1 | - | 1 - 2 | 255 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
