Assay Details
| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503319.2 | XP_003503367.1 | 7 - 8 | 1639 | 98 | |
| GenBank mRNA | JI884734.1 | - | 2 - 3 | 173 | 98 | |
| JP044670.1 | - | 6 - 7 | 1673 | 98 | ||
More Information
Set Membership: |
|

Agpat6
Genomic Map