Assay Details
Gene Symbol: Arl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510923.2 | XP_003510971.1 | 5 - 6 | 668 | 64 | |
GenBank mRNA | JI891227.1 | - | 5 - 6 | 538 | 64 | |
JP051985.1 | - | 5 - 6 | 538 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
