Assay Details
Gene Symbol: LOC100753059 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648757.1 | XP_007646947.1 | 79 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007648757.1 transcripts.
Set Membership: |
![]() |
