Assay Details
| Gene Symbol: Slc22a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512210.2 | XP_003512258.1 | 9 - 10 | 1576 | 68 | |
| GenBank mRNA | JI884485.1 | - | 5 - 6 | 725 | 68 | |
| JP061098.1 | - | 5 - 6 | 725 | 68 | ||
More Information
Set Membership: |
|

Slc22a5
Genomic Map