Assay Details
Gene Symbol: Crabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502904.2 | XP_003502952.1 | 1 - 2 | 67 | 66 | |
XM_007643519.1 | XP_007641709.1 | 1 - 2 | 67 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
