Assay Details
| Gene Symbol: Ube2l6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506156.2 | XP_003506204.1 | 3 - 4 | 370 | 74 | |
| XM_007646636.1 | XP_007644826.1 | 2 - 3 | 274 | 74 | ||
More Information
Set Membership: |
|

Ube2l6
Genomic Map