Assay Details
Gene Symbol: Tbccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508224.2 | XP_003508272.1 | 3 - 4 | 869 | 61 | |
XM_007648846.1 | XP_007647036.1 | 3 - 4 | 651 | 61 | ||
XM_007648847.1 | XP_007647037.1 | 3 - 4 | 604 | 61 | ||
XM_007648848.1 | XP_007647038.1 | 3 - 4 | 679 | 61 | ||
GenBank mRNA | JP057928.1 | - | 3 - 4 | 835 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
