Assay Details
| Gene Symbol: LOC100769195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643082.1 | XP_007641272.1 | 23 - 24 | 2881 | 81 | |
| GenBank mRNA | JP045526.1 | - | 22 - 23 | 2874 | 81 | |
More Information
Set Membership: |
|

LOC100769195
Genomic Map